Learn More
Bohlender™ SICCO™ Mini Vitrum Desiccator
Mini Vitrum Desiccator made of borosilicate glass 3.3 with aluminum frame.
Brand: Bohlender™ V1811-07
This item is not returnable.
View return policy
Description
- Aluminum frame with panels made of heat-resistant borosilicate glass 3.3
- Includes one shelf made of aluminum, hygrometer and desiccant
- Usable with up to one shelf, stackable
Specifications
Borosilicate Glass 3.3 | |
1.6 gal. | |
9 in. | |
7.2 in. | |
8.4 in. | |
4.4 lb. |
Silica Gel Jar (23 g) | |
6 L | |
220 mm | |
183 mm | |
214 mm | |
2 kg |