Learn More
Bohlender™ SICCO™ Mini Vitrum Desiccator
Mini Vitrum Desiccator made of borosilicate glass 3.3 with aluminum frame.
Brand:  Bohlender™ V1811-07
Product Code. 17157694
  • € 1770.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Description
  • Aluminum frame with panels made of heat-resistant borosilicate glass 3.3
  • Includes one shelf made of aluminum, hygrometer and desiccant
  • Usable with up to one shelf, stackable
Specifications
Specifications
Borosilicate Glass 3.3
1.6 gal.
9 in.
7.2 in.
8.4 in.
4.4 lb.
Silica Gel Jar (23 g)
6 L
220 mm
183 mm
214 mm
2 kg