missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ SICCO™ Mini Desiccators Black Premium

Mini Desiccators Black Premium made of black polycarbonate.

Brand:  Bohlender™ V1940-06

Product Code. 17117734

  • € 349.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Black polycarbonate prevents light incidence and protects against ultraviolet radiation
  • Includes two shelves, hygrometer and desiccant, non-slip rubber feet, stackable
  • One Touch Door with circular seal
  • Robust construction
  • Shatterproof cabinet made of polycarbonate
  • Hinge door with wide opening angle
  • Lightweight, transportable
  • Stackable
  • Capacity: 6.2 L
  • Desiccant in a handy, resealable container included in delivery
Specifications

Specifications

Polycarbonate
2 Shelves
221 mm
183 mm
214 mm
0.9 kg
Silica Gel Jar (23 g)
8.7 in.
7.2 in.
8.5 in.
2 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers