Learn More
Honeywell Safety Products™ Shoe, safety, Bacou Mont Blanc, white, S2 T37, Sperian
Product Code. 12691293
Product Code. 12691293
Brand: Honeywell Safety Products™ 623321637/7
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mont Blanc safety shoes.
Mocassin slip-on leather shoe with limited stitching on the upper. Lined and padded collar. Anti-puncture midsole. Metallic toe cap 200J, perforated insole, polyurethane white with double density injected outsole.European Directive: 89/686/EEC - EC PPE Category II - Standard: EN 345-1 - Marking : S2/S3 Quality Assurance: ISO 9001:2008.
  • Size 37
  • Pair