Learn More
Justrite Shelves for EN Cabinet 226-EN 2903
Product Code. 17792388
Click to view available options
Model:
JCBS120S00
JCBS60S000
JCBSROLL00
Length (Metric):
49 cm
109 cm
Unit Size:
Each
Product Code. 17792388
Brand: Justrite JCBS60S000
Additional shelf accessory for EN Cabinets 226-EN. Choice of installed or pull out shelves in stainless steel or epoxy coated finishings.
  • Choice of stainless steel or epoxy coated shelves
  • Choice of installed or pull out shelves
Specifications
For Use With (Equipment) EN Cabinet 226-EN
Length (Metric) 49 cm
Model JCBS60S000
Width (Metric) 42 cm
Color Light Gray
Material Steel
Product Type Shelf