missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Mouse IgG whole molecule, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB9380.2mg
This item is not returnable.
View return policy
Specifications
Sheep anti-Mouse IgG whole molecule | |
Polyclonal | |
Sheep anti-mouse IgG recognizes mouse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. | |
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Mouse IgG whole molecule | |
2 mg | |
Mouse | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Sheep | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only