Learn More
SharpSafe™ Polypropylene Containers
Product Code. p-8009820
Click to view available options
Capacity (Metric):
0.3 L
0.45 L
Height (Metric):
130 mm
169 mm
Unit Size:
Each
Pack of 100
Product Code. 11969157
Brand: SharpSafe™ SHA4175
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use yellow SharpSafe™ Polypropylene Containers for the collection of sharp waste treatment activities and instruments that are pungent, sharp and/or pointed.
  • Translucent flip-top lid and handle
  • Yellow
  • Polypropylene
  • Autoclavable
Specifications
Capacity (Metric) 0.3 L
Certifications/Compliance Complies with UN 3291 and BS 7320 standard, NF-X-30–500, NF302, A.D.R. and UN3291, TRBA 250, Quality management system: ISO 9001:2008 and ISO 13485:2003, Environmental Standard: ISO 14001:2004
Closure Type Flip Top
Closure Color Translucent
Color Yellow
Dimensions (L x W x H) 105 x 50 x 130 mm
Height (Metric) 130 mm
Length (Metric) 105 mm
Opening Size (Metric) 55 x 50 mm
Quantity 100 Pk.
Width (Metric) 50 mm
Autoclavable Autoclavable
Includes Flip top lid, handle
Material Polypropylene