Learn More
SharpSafe™ Polypropylene Containers
Click to view available options
Capacity (Metric):
0.3 L
0.45 L
Height (Metric):
130 mm
169 mm
Unit Size:
Each
Pack of 100
Description
- Translucent flip-top lid and handle
- Yellow
- Polypropylene
- Autoclavable
Specifications
Specifications
| Capacity (Metric) | 0.3 L |
| Certifications/Compliance | Complies with UN 3291 and BS 7320 standard, NF-X-30–500, NF302, A.D.R. and UN3291, TRBA 250, Quality management system: ISO 9001:2008 and ISO 13485:2003, Environmental Standard: ISO 14001:2004 |
| Closure Type | Flip Top |
| Closure Color | Translucent |
| Color | Yellow |
| Dimensions (L x W x H) | 105 x 50 x 130 mm |
| Height (Metric) | 130 mm |
| Length (Metric) | 105 mm |
| Opening Size (Metric) | 55 x 50 mm |
| Quantity | 100 Pk. |
| Show More |