Learn More
Epredia™ Shandon™ Para-Release™ Spray
Ease the release of paraffin blocks from embedding molds with Epredia™ Shandon™ Para-Release™ Spray.
Brand:  Epredia™ 9990420
Product Code. 11306800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Apply spray-releasing agent to molds before adding paraffin for clean release of block.
Specifications
Specifications
50 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers