Learn More
Epredia™ Shandon™ EA-50 Stain
Clearly differentiate cellular components with pink, green and blue-green hues utilizing Epredia™ Shandon™ EA-50 Stain.
Brand:  Epredia™ 6766001
 View more versions of this product
Product Code. 10167988
  • € 77.95 / Pack of 2
Explore more special offers
Description
Description
  • Polychromatic cytoplasmic stain
  • Biological stain commission-certified dye
Specifications
Specifications
1L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Cytological stain EA-50 / EA-65
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol, Isopropylalcohol, Phosphotungstic acid, Eosin-YDye