Learn More
Epredia™ Cryomatrix™ embedding resin
Secure and support tissue with Epredia™ Cryomatrix™ embedding resin, an outstanding product used to support tissue for frozen sectioning.
Brand:  Epredia™ 6769006
Product Code. 12868290
Description
Description
  • Forms a firm bond with the specimen holder
  • Freezes quickly, sections smoothly
  • Dissolves in water without residue
TRUSTED_SUSTAINABILITY
Specifications
Specifications
120 mL
Embedding Media
Product Suggestions
Product Suggestions
Videos
SDS Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Cryomatrix™ embedding resin > Cryomatrix embedding resin