Learn More
Trajan™ SolGel WAX Columns
Polyethylene Glycol (PEG) in a Sol-Gel matrix. Robust, high temperature polar WAX phase
Brand: Trajan™ 054788
This item is not returnable.
View return policy
Description
- Bonded polyethylene glycol
- Very robust high-temperature column
- Less susceptible to damage by oxygen than conventional wax phases
- Polar phase
- Low bleed and inert
Specifications
280°C | |
0.32 mm | |
G14, G15, G16, G20, G39 | |
30°C to 260/280°C | |
30 m | |
Polar |
Hydrogen Bonding or Dipole interactions | |
Polyethylene Glycol | |
Essential Oils Analysis, Amines Aliphatic, Amines Aromatic, Alcoholic Beverages Analysis, Essential Oils, Food - FAME, Phenols, Sugars-alditol Acetates | |
0.25 μm | |
SolGel-Wax | |
Fused Silica |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Trajan™ SolGel WAX Columns