Learn More
Trajan™ Replacement Valves for Luer Lock Syringes
Click to view available options
For Use With (Equipment):
Syringe (5 mL to 2 L)
Syringe (50μL to 2 L)
Unit Size:
Each
Description
- Push-pull valve with Luer Lock termination to suit removable needle 25μL-2.5mL syringes
- Push button valve with Luer Lock termination. Suits 5mL-2L syringes with removable Luer Lock or removable needle
- Luer Lock valve for any Luer Lock and Luer Tip syringes or other Luer devices. Ideal when fast needle changeover is needed
Specifications
Specifications
| Description | Push buttone with Luer Lock |
| Product Type | Replacement Valve |
| For Use With (Equipment) | Syringe (5 mL to 2 L) |