Learn More
Trajan™ PerkinElmer Syringe
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
0.5 μL
missing translation for 'unitSize'
Each
Description
- Specifically designed for PerkinElmer instruments
Specifications
Specifications
| Needle Gauge | 26 G |
| Diameter (Metric) Inner Needle | 0.155 mm |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Volume (Metric) | 0.5 μL |
| Needle Point Style | Beveled Cone |