Learn More
SGE™ MEPS™ Syringes
Product Code. 12917596
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
For Use With (Equipment):
CTC Analytics
CTC Analytics, HTA 300APlus, Thermo Scientific and Varian 8400 System
HTA 300APlus, Thermo Scientific and Varian 8400 System
eVol Automated Syringe
Product Type:
MEPS™ Syringe
Syringe
Unit Size:
Each
Product Code. 12917596
Brand: SGE™ 2910026
For use in micro-SPE
  • The eVol™ custom programming function enables MEPS to be semi-automated and proofed before transition to fully automated platforms
Specifications
For Use With (Application) Gas/Liquid chromatography
Product Type Syringe
Quantity Ea.
Volume (Metric) 500 μL
For Use With (Equipment) eVol Automated Syringe