Learn More
Trajan™ eVol™ XCHANGE Syringes
Click to view available options
Volume (Metric):
100 μL
1 mL
Quantity:
1 Pk.
3/Pk.
Unit Size:
Each
Pack of 3
Description
- Liquid handling user independent device
- Reduction in need for re-analysis
- Syringes easily and quickly changed
- Replacement parts available
Specifications
Specifications
| Needle Gauge | 23 G |
| For Use With (Application) | Liquid handling procedure |
| Length (English) | 1.9 in. |
| Length (Metric) | 50 mm |
| Product Type | eVol™ Syringe |
| Quantity | 1 Pk. |
| Volume (Metric) | 1 mL |
| Length (Metric) Needle | 50 mm |
| Needle Point Style | Bevel |