Learn More
Trajan™ Diamond Thermo RSH 10 to 50μL GC Autosampler Syringes
Click to view available options
Diameter (Metric) Inner Needle:
0.11 mm
0.24 mm
Includes:
Ultra Smooth Coating
Ultra Smooth Hydrophilic Coating
Ultra Smooth Hydrophobic Coating
Unit Size:
Each
Description
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
| Description | Needle coating: Ultra Smooth |
| Needle Gauge | 23 G |
| Diameter (English) Inner Needle | 0.009 in. |
| Diameter (Metric) Inner Needle | 0.24 mm |
| Diameter (English) Outer Needle | 0.024 in. |
| Diameter (Metric) Outer Needle | 0.63 mm |
| For Use With (Application) | Gas chromatography |
| Includes | Ultra Smooth Coating |
| Length (English) Needle | 2.24 in. |
| Product Type | GC Autosampler Syringe |
| Show More |