Learn More
Trajan™ Autosampler Syringes
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
10 μL
Needle Gauge:
23 G
25 G
26 G
Connection Type:
Fixed
Plunger-In, Repeating Adapter
Removable
For Use With (Equipment):
Carlo Erba/Fisons/Leap/CTC AS200 and AS800, GC autosampler syringe
Carlo Erba/Fisons/Leap/CTC ASD200 and AS80, triPlus
PerkinElmer Autosystem
Shimadzu AOC 14, 17, 20 and 20i GCs
Varian 8035, 8100 and 8200 GC Autosamplers
Unit Size:
Each
Description
Replace syringes for Agilent™ Technologies, Varian™, Perkin Elmer™, Shimadzu™, Carlo Erba™, Fisons™, CTC/Leap™, Hitachi™, and more
Syringe needles are cone point unless otherwise noted.
Order Info
Additional syringes are available for other autosamplers made by the manufacturers represented here, as well as syringes for Unicam™, Waters™, and Siemens™ instruments. For information, contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
| Needle Gauge | 26 G |
| Diameter (English) Outer Needle | 0.01 in. |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Length (English) | 2 in. |
| Length (English) Needle | 1.9 in. |
| Length (Metric) | 51 mm |
| Product Type | Autosampler Syringe |
| Model | 5F-C/T-5/0.47C |
| Quantity | Ea. |
| Volume (Metric) | 5 μL |
| Show More |