Learn More
Trajan Scientific and Medical™ SGE Analytical Science Quartz Wool
GC inlet liner for use with Pyrojector II and pyrolysis chromatography.
Brand:  Trajan Scientific and Medical™ 18060001
Product Code. 15259193
  • € 70.95 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Description
  • High Resolution Capillary GC
  • For use with Pyrolysis chromatography applications
  • Works in conjunction with Pyrojector II which thermally fragments essentially non-volatile molecules into volatile fragments, making them suitable for chromatographic analysis
Specifications
Specifications
GC Inlet Liner
Quartz
Quartz Wool
0.5 g
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Trajan Scientific and Medical™ SGE Analytical Science Quartz Wool > Weight: 0.5g