Learn More
Cytiva SFX-Insect™ Cell Culture Media
Product Code. p-7058053
Click to view available options
Quantity:
1000 mL
5 L
10 L
6 x 1000 mL
With Additives:
Glutamine, Poloxamer 188
Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Protein, No Phenol Red, No HEPES
No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES
Form:
Liquid
Powder
Unit Size:
10 L
1000mL
5 L
Pack of 6
Product Code. 10024393
Brand: Cytiva SH30278.LS
Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.
  • Protein free media
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type Insect Cell Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives No Protein, No Phenol Red, No HEPES
Quantity 6 x 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light