Learn More
Thermo Scientific™ Sensititre™ Equine EQUIN1F Vet AST Plate
Perform accurate AST with an equine-specific formulary with this convenient plate.
Brand: Thermo Scientific™ EQUIN1F
Description
Perform accurate AST with an equine-specific formulary with the Thermo Scientific™ Sensititre™ Equine EQUIN1F AST Plate.
Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Equine EQUIN1F AST Plate.
- Extended shelf life, room temperature storage eliminates inventory control concerns.
- Individual packaging allows the laboratory to test one plate at a time with no waste.
- Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
- Automated or manual read options are available for ultimate flexibility.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
Specifications
Various | |
Various | |
Non-fastidious, Gram-positive and Gram-negative Veterinary Isolates | |
1 | |
Autoread or manual |
Sensititre Equine MIC Plate | |
AST Plate | |
Veterinary AST, Minimum Inhibitory Concentration (MIC) | |
10/Pk. | |
Equine Isolates |