missing translation for 'onlineSavingsMsg'
Learn More
Semicarbazide hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 11409153

Semicarbazide hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11409153
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11409153

Brand: Thermo Scientific Alfa Aesar A11668.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Urease substrate and MAO inhibitor

Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Semicarbazide hydrochloride is used as urease substrate and MAO inhibitor. Derivatizing agent for carbonyl compounds as their semicarbazones which produces crystalline compounds with characteristic melting points. Also used in heterocyclic synthesis. Semicarbazone formation has been used to separate carbonyl compounds from mixtures by adsorption onto silica gel from a hydrocarbon solution. Regeneration is by hydrolysis with oxalic acid.

Solubility
Soluble in water (>100g/L).

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-41-7
Molecular Formula CH6ClN3O
Molecular Weight (g/mol) 111.53
MDL Number MFCD00013009
InChI Key XHQYBDSXTDXSHY-UHFFFAOYSA-N
Synonym semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochlorideShow MoreShow Less
PubChem CID 11236
ChEBI CHEBI:82532
IUPAC Name aminourea;hydrochloride
SMILES [H+].[Cl-].NNC(N)=O

Specifications

Melting Point ∼176°C (decomposition)
pH 1.5
Odor Odorless
Linear Formula H2NCONHNH·HCl
Quantity 2500 g
UN Number UN2811
Beilstein 3593642
Merck Index 14,8444
Solubility Information Soluble in water (>100g/L).
Formula Weight 111.53
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Semicarbazide hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253025 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberA11668.22,A11668.36,A11668.0E
Lot Number10221203 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog NumberA11668.22,A11668.36,A11668.0E
Lot Number10223275 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog NumberA11668.22,A11668.36,A11668.0E
Lot Number10202666 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog NumberA11668.22,A11668.36,A11668.0E
Lot NumberJ4470A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11668.22,A11668.36,A11668.0E
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H318-Causes serious eye damage.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Semicarbazide hydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.