Learn More
VITLAB™ Sedimentation Cone
Manufactured according to DIN 12 672. VITLAB™ Sedimentation Cone is used to determine suspended matter in liquids.
Brand:  VITLAB™ 75991
Product Code. 11774129
  • € 110.00 / Pack of 3
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear
  • With raised scale 
  • Screw coupling can be removed
Order Info
CapacityMetric
Specifications
Specifications
1000 mL
SAN
3
Sedimentation Cone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers