Learn More
DWK Life Sciences Sealing ring, for polished spherical joints (silicone + FEP)
Product Code. 15807091 Shop All DWK Life Sciences Products
Click to view available options
Description:
19 NS
29 NS
35 NS
41 NS
64 NS
S 13 NS
Unit Size:
Pack of 10
Product Code. 15807091
Brand: DWK Life Sciences 286236402
Only null left
Add to basket
Add to basket
For sealing DURAN™ ball joints without ground joints with high demands on chemical resistance.
Non-sanded DURAN™ spherical ground joints only seal via the sealing ring. FEP-coated silicone rings are used where high chemical resistance is required.
  • Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
  • Silicone and FEP sealing ring
TRUSTED_SUSTAINABILITY
Specifications
Description 64 NS
Autoclavable Not Autoclavable
Material Silicone, FEP
For Use With (Application) Sealing of non-sanded spherical ground joints
Packaging Quantity 10/Pk.