Description
This easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™, JOE™, NED™, ROX™, SYBR™ Green, VIC™, TAMRA™, and TET™ dyes on the ABI PRISM 7700™ Sequence Detection System. The SDS Spectral Calibration kit includes 8 tubes of pure dye standards for the Applied Biosystems™ ABI PRISM 7700™ Sequence Detection System. Spend Your Time on Research - Not Instrument Calibration This kit enables users to calibrate the ABI PRISM 7700™ Sequence Detection System quickly and easily. The system automatically collects the dye standard's spectral information as part of the instrument installation procedure. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
Specifications
Specifications
| Content And Storage | Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions. Includes 8 tubes of pure dye standards (FAM™, JOE™, NED™,ROX™, SYBR™ Green, VIC™, TAMRA™, TET™), for ABI PRISM™ 7700 Sequence Detection System. |
| For Use With (Equipment) | 7700 System |
| Product Type | Spectral Calibration Kit |
| For Use With (Application) | Real time PCR (qPCR) |
| Description | Tubes |
| Quantity | 8 Tubes |
| Calibration Type | Spectral Calibration |