Learn More
Labconco™ Scroll Vacuum Pump
For use with FreeZone™ Freeze Dry Systems - Oil-free, dry pumps requiring very little maintenance. Labconco™ Scroll Vacuum Pump is dry pump that use two spiral scrolls to compress air and vapors and move them toward exhaust.
Brand:  Labconco™ 7587002
 View more versions of this product
Product Code. 15914232
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Scroll recommended be changed every 40,000 hours of use
  • Handle water vapor better than most types of pumps and make significantly less noise during operation
  • Displacement capacity of 189 to 195L/min., dependng on the Hz of operation
  • Completely oil-free pumps with true hydrocarbon-free design
  • All models feature acid-resistant Chemraz™ valves and stainless steel fittings for corrosion-resistance
  • Have an inverter so voltage operation may be changed in field if needed
  • Vacuum outlet is 1.9cm (0.75 in.) OD
  • May be used with aqueous and solvent samples including acetonitrile, as well as concentration applications
Specifications
Specifications
230 V 50/60 Hz
61 lb.
189 L/min.
15 in.
12 in.
50.8 cm
28 kg
FreeZone Freeze Dry Systems
38.1 cm
30.4 cm