Learn More
Alpha Packaging™ Screw Cap
Product Code. 15458964
Click to view available options
Material:
Bakelite
Bakelite screw cap without liner
Specifications
Material Bakelite
Certifications/Compliance DIN 45
Closure Liner Unlined
Closure Size 617