Learn More
Berlin Packaging™ Phenol Black Bottle Closures
Product Code. 30408797
Click to view available options
Certifications/Compliance:
DIN
DIN 18
DIN 55
Unit Size:
Each
Pack of 750
Product Code. 30408797
Brand: Berlin Packaging™ 1000629
DIN certified.
  • DIN certified
  • Screw top
  • For glass bottles with narrow neck
  • Black
Specifications
Color Black
Material Phenolic Resin
Closure Color Black
Certifications/Compliance DIN 55
Shape Round
Product Type Screw Cap