Learn More
Alpha Packaging™ Phenol Black Bottle Closures
Click to view available options
Certifications/Compliance:
DIN
DIN 18
DIN 55
Description
- DIN certified
- Screw top
- For glass bottles with narrow neck
- Black
Specifications
Specifications
Color | Black |
Material | Phenolic Resin |
Closure Color | Black |
Certifications/Compliance | DIN |
Shape | Round |
Product Type | Closure |
Product Content Correction
Product Title
Alpha Packaging™ Phenol Black Bottle Closures >