Learn More
Porvair Sciences™ Science Polystyrene White Microplate
Brand:  Porvair Sciences™ 222003
Encompass_Preferred
Product Code. 12469327
  • € 786.10 / Pack of 100
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Square wells maximise liquid volume per well
  • White plate prevents cross talk in light emitting assays
  • Black plate gives low autofluorescence background
  • Volume, total μL/well: 120
  • Format: 24 x 16 wells
  • Dimensions [w x d x h], mm: 3.75 x 3.75 x 11.4 (well); 85.5 x 127.5 x 14.8 (plate)
Specifications
Specifications
Standard
White
Polystyrene
120 μL
100 Each
16 x 24
Microplate
384
100
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Porvair Sciences™ Science Polystyrene White Microplate