Learn More
Thermo Scientific™ Schaedler Anaerobe Agar / Schaedler Anaerobe KV Selective Agar Biplate
Grow obligate and facultative anaerobic organisms and isolate anaerobic Gram-negative bacteria, Bacteroides and Prevotella species, on highly nutritive biplates.
Brand: Thermo Scientific™ PB5204E
18.61 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Grow obligate and facultative anaerobic organisms and selectively isolate anaerobic Gram-negative bacteria, especially Bacteroides and Prevotella species, with Thermo Scientific™ Schaedler Anaerobe Agar / Schaedler Anaerobe KV Selective Agar biplates. The highly nutritive media create selective conditions for the delicate and more nutritionally exacting microorganisms of the intestinal tract. The convenient biplate format saves space in incubators and kits for gas-generating atmosphere.
Conveniently isolate fastidious microorganisms of the intestinal tract, which would normally be swamped by the growth of enterococci, coliform bacilli and other Gram-negative bacilli, with Schaedler Anaerobe Agar / Schaedler Anaerobe KV Selective Agar biplates.
- Highly nutritive for the selective isolation of intestinal bacteria
- Ready-to-use convenience of prepared media
- Save space with the biplate format
Schaedler Anaerobe Agar is a medium free from thioglycollate for the growth of aerobic and anaerobic organisms. Schaedler, Dubos and Costello1 formulated this medium for the isolation of aerobic and anaerobic microorganisms from the gastro-intestinal tract of mice. Mata, Carrillo and Villatoro2 modified the formula in their studies on anaerobic human faecal microflora.
Schaedler Anaerobe Agar contains cysteine hydrochloride and glucose, as reducing substances, with the advantage that cysteine inhibits the growth of Escherichia coli. Schaedler Anaerobe Agar has been shown to be a suitable alternative to blood agar for the enumeration of clostridia and has been used for the examination of food, waste products and ditch water. Studies have shown the necessity for strict anaerobic conditions for the successful recovery of obligate anaerobes when using this medium without the addition of blood.
Schaedler Anaerobe KV Selective Agar is supplemented with kanamycin and vancomycin. Together these select for anaerobic Gram-negative bacteria, especially Bacteroides and Prevotella species.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2° to 12°C, in the dark | |
90mm biplate | |
17.5g ± 5% | |
10/Pk. |
Schaedler Anaerobe Agar / Schaedler Anaerobe KV Selective Agar | |
Agar | |
Wrapped in single cellulose-based film wrap |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6249111 | Certificate of Analysis | 24/03/2025 | PB5204E |
6249110 | Certificate of Analysis | 24/03/2025 | PB5204E |
6245921 | Certificate of Analysis | 17/03/2025 | PB5204E |
6245920 | Certificate of Analysis | 17/03/2025 | PB5204E |
6242421 | Certificate of Analysis | 10/03/2025 | PB5204E |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.