Learn More
Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished two sides, C-axis, HEMCOR single crystal
€ 267.33 - € 1157.76
Specifications
Product Type | Sapphire Substrate, EPI Polished Two Sides |
---|---|
Description | EPI polished two sides |
Diameter (Metric) | 50.8 mm |
Specimen Size | 50.8mm dia. x 0.332 mm thick |
Thickness | 0.332 mm |
Product Code | Brand | Quantity | Specimen Size | Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Specimen Size | Thickness | Price | Quantity & Availability | |||
15463925
|
Thermo Scientific Chemicals
045020.KT |
1 Ea. | 50.8mm dia. x 0.332 mm thick | 0.332 mm |
€ 267.33
Each |
In Stock
Log in to see stock. |
|||
15473925
|
Thermo Scientific Chemicals
045020.KU |
5 Ea. | 50.8mm dia. x 0.332 mm thick | 0.332 mm |
€ 1157.76
Pack of 5 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sapphire Substrate, EPI Polished Two Sides | |
50.8 mm | |
0.332 mm |
EPI polished two sides | |
50.8mm dia. x 0.332 mm thick |