Learn More
Buerkle™ Samplers Transport Case
Buerkle™ Samplers Transport Case can be used for Buerkle™ Samplers.
€ 20.60 - € 164.00
Specifications
Product Type | Transport Case |
---|---|
Color | Black |
For Use With (Equipment) | For Samplers |
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
10474982
|
Buerkle™
3600-2120 |
123 x 25 x 11 cm |
€ 164.00
Each |
In Stock
Log in to see stock. |
|||||
10522743
|
Buerkle™
3600-0021 |
265 x 225 x 50 mm |
€ 20.60
Each |
In Stock
Log in to see stock. |
|||||
Description
- For use with samplers
Specifications
Transport Case | |
Black | |
For Samplers |