Learn More
Thermo Scientific™ Sample and Reagent Cups for iCE™ 3000 Series AA systems
Ensure consistent sample presentation and complete sample pick-up with sample and reagent cups for Thermo Scientific™ iCE™ 3000 Series AA systems.
Brand:  Thermo Scientific™ 942339380031
 View more versions of this product
Product Code. 10461240
  • € 282.00 / Pack of 1000
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Description
Use Thermo Scientific™ Sample and Reagent Cups with Thermo Scientific™ GFS33 and GFS35(Z) Graphite Furnace Autosamplers to ensure consistent sample presentation and complete sample pick-up. Select from polypropylene sample and reagent cups, PTFE sample cups, and reduced volume sample cups.
The perfect choice for the Thermo Scientific™ GFS33 and GFS35(Z) Graphite Furnace Autosamplers, when used with our iCE 3000 Series Atomic Absorption spectrometers; these Thermo Scientific Sample and Reagent Cups for Thermo Scientific™ iCE™ 3000 Series AA systems ensure consistent sample presentation and complete sample pick-up. Select from polypropylene sample and reagent cups, PTFE sample cups, and reduced volume sample cups, according to your needs.
Specifications
Specifications
GFS33 and GFS35(Z) Graphite Furnace Autosamplers
Polypropylene Sample Cups
Polypropylene Sample Cups for iCE™ 3000 Series AA Systems
1000/Pk.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Sample and Reagent Cups for iCE™ 3000 Series AA systems >