Learn More
Saint Gobain Life Sciences Chemware™ Raschig Rings
Click to view available options
Diameter (Metric) Outside:
3 mm
6 mm
Length (Metric):
3 mm
6 mm
Unit Size:
Pack of 300
Pack of 500
Description
- Packing for distillation columns and scrubbers.
- Chemically inert except for molten alkali metals, fluorine, and strong fluorinating agents.
- High surface area per unit of volume.
- Minimum resistance to gas and liquid flow.
Specifications
Specifications
| For Use With (Application) | For chemical processes and liquid or gas transfer applications |
| Length (Metric) | 3 mm |
| Type | Raschig Ring |
| Material | PTFE |
| For Use With (Equipment) | For distillation columns |
| Diameter (Metric) Outside | 3 mm |
| Length (English) | 0.1 in. |
| Diameter (English) Outside | 0.1 in. |