Learn More
Saint Gobain Life Sciences Bytac™ Surface Protector
Click to view available options
Dimensions (L x W):
15 ft. x 12.5 in.
15 ft. x 25 in.
Material:
FEP
FEP, Aluminum
FEP, Vinyl
Unit Size:
4.5m
4.6m
5m
Each
Pack of 5
Beschreibung
Put the protection of FEP fluoropolymer on any surface
- Made of an overlay of FEP film bonded to a support backing of vinyl or aluminum (backing is coated with pressure-sensitive acrylic adhesive)
- Protect benchtops, fume hoods, temporary tables, walls/ceilings, splash screens, walls of spray booths, and other lab surfaces
- Help protect surfaces against absorption of toxic or radioactive chemicals
- Corrosive chemicals, paint, oil, grease, and sticky substances wipe off easily with damp sponge
- Cut easily with scissors
- Install easily—simply peel off protective paper from adhesive backing and apply to surface
Bestellinformation
Bytac protector with vinyl backing recommended for temperatures up to 93°C (200°F); with aluminum foil backing, can be used up to 182°C (360°F).
Spezifikation
Spezifikation
| Product Type | Bench Protector |
| Material | FEP, Aluminum |
| Length (English) | 15 ft. |
| Length (Metric) | 4.6 m |
| Width (English) | 25 in. |
| Width (Metric) | 640 mm |
| Dimensions (L x W) | 15 ft. x 25 in. |
| For Use With (Application) | Protects benchtops, fume hoods, temporary tables, walls/ceilings, splash screens, walls of spray booths, and other lab surfaces |
| Quantity | 1 Ea. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur