Learn More
Deutsch Neumann™ Safety Gas Hoses
Click to view available options
Length (Metric):
500 mm
600 mm
750 mm
1000 mm
1250 mm
Unit Size:
1250mm
1500mm
500mm
600mm
Each
Description
- Working pressure to 100 mbar
- For gas burners meeting DIN 30 665
- For all gases meeting DVGW Work Code G 260
Specifications
Specifications
| Product Type | Gas Hose |
| For Use With (Equipment) | Gas burners meeting DIN 30 665 and all gases |
| Length (Metric) | 1250 mm |