missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Safety Equipment Zero Waste Box™

Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Description
Use this Zero Waste Box to recycle:
- Earplugs
- Beard nets
- Hairnets
- Disposable gloves
- Safety glasses
- Disposable garments
- Disposable shoe covers
- Disposable masks*
*We accept all types of disposable face masks made from plastic, such as dust masks, FFP2 (or so-called ''N95''), FFP3 and surgical masks (as long as they don't come from a medical environment).
Do not send in:
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Autoclaved material
- Fabric, homemade or paper face masks
- Wax earplugs
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which present a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass
Specifications
Specifications
| Capacity (Metric) | 15 kg |
| Capacity (English) | 33 lb. |
| Color | Beige, Green, Red, White |
| Dimensions (L x W x H) | 30.5 x 30.5 x 84 cm |
| Disposable | Single-use |
| Latex Free | Yes |
| Size | Medium |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Cardboard, Plastic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction