Learn More
Justrite™ Safety Can For Liquid Disposal
Product Code. p-7070903
Click to view available options
Capacity (English):
2 gal.
5 gal.
Color:
Red
White
Height (English):
14.8 in.
20 in.
Unit Size:
Each
Product Code. 12923695
Brand: Justrite™ 12772
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Safety Can For Liquid Disposal provides a better solution for the containment of liquid flammable or corrosive waste.
High-density polyethylene (HDPE) disposal cans provide superior chemical resistance and leak protection. Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig (0.2 and 0.35 bar). Stainless steel handle and hardware with latch-catch system hold the cap in the open position for easy filling and pouring. Large-mouth opening features a built-in stainless steel flame arrester to guard against outside ignition. The broad-base design provides stability and resists accidental tip over.
Specifications
Product Type Safety Can
Description Safety Can for Liquid Disposal
Capacity (English) 5 gal.
Color White
For Use With (Application) Liquid Disposal
Material HDPE
Includes Stainless steel handle, faucet and flame arrester
Height (English) 20 in.