Learn More
Heidolph Instruments S-Box Package 2
The S-Box Package 2 includes additional accessories to complete an existing S-Biosystem incubation system.
Brand: Heidolph Instruments 400-03001-00
Description
- Each S-box is a stand-alone miniature incubator.
- Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
- The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
- Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
- To avoid mix-up, the front cover of the S-box is available in different colors.
Specifications
Orange | |
1 S-box, 1 tool kit, 8 inner boxes, 8 water bags, 4 front covers |