Learn More
Eppendorf™ Rubber Mats
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
500mL rectangular bucket
Adapter 5702724009
Adapter 5702735000
Adapter 5702737003
Adapters for Rotor A-4-44
Rotor A-8-17
Rotor F-35-30-17
Capacity:
1 x 10 mL Vessel
1 x 15 mL Round Bottom Tube
1 x 15 mL Vessel
1 x 500 mL Bucket
1 x 7 mL Vessel
4 x 100 mL Tubes
Unit Size:
Each
Pack of 10
Pack of 16
Pack of 20
Pack of 4
Pack of 8
Description
- Rubber mats
- Available for a variety of different rotors and adapters
Specifications
Specifications
| Type | Mat |
| Material | Rubber |
| For Use With (Equipment) | Adapter 5702735000 |
| Color | Black |
| Capacity | 1 x 10 mL Vessel |
| Includes | Set of 20 mats |