Learn More
OHAUS™ Rubber Mat
Product Code. 15762479 Shop All Ohaus Products
Click to view available options
Dimensions (L x W):
28 x 33 cm
33 x 33 cm
45.7 x 45.7 cm
48 x 61 cm
61 x 61 cm
61 x 91.4 cm
Unit Size:
Each
Product Code. 15762479
Brand: OHAUS™ 30400064
Only null left
Add to basket
Add to basket
OHAUS™ Rubber Mat is an accessory for use with various platforms.
  • For use with platforms
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
  • Securely hold labware in place
TRUSTED_SUSTAINABILITY
Specifications
Product Type Platform Mat
Material Rubber
Depth (English) 24.00 in.
Depth (Metric) 61.00 cm
Dimensions (L x W) 61 x 61 cm
Length (English) 24.00 in.
Length (Metric) 61.00 cm
Width (English) 24.00 in.
Width (Metric) 61.00 cm
Description Rubber Mat, 61 X 61 cm
For Use With (Equipment) Platform