Learn More
Fisherbrand™ Rubber Cone Filter Crucible Adapters
Shop All Fisherbrand Products
Click to view available options
Cone Size:
27 mm I.D., 18 mm I.D.
34 mm I.D., 23 mm I.D.
40 mm I.D., 25 mm I.D.
40 mm I.D., 27 mm I.D.
50 mm I.D., 30 mm I.D.
60 mm I.D., 40 mm I.D.
Height (Metric):
25mm
30mm
35mm
45mm
50mm
55mm
Unit Size:
Pack of 10
Specifications
Specifications
| Cone Size | 34 mm I.D., 23 mm I.D. |
| For Use With (Equipment) | Filter crucibles |
| Height (Metric) | 30mm |
| Material | Rubber |
| Product Type | Cone |
| Quantity | 10Pack |