Learn More
Abeba™ Rubber 9300 Shoes
Acid and base resistant. Rubber sole provides high slip resistance.
Brand: Abeba™ 9300/47
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- White
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Leather insole
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li | |
Leather | |
Slip-resistant | |
9300 |
White | |
47 | |
Rubber |