missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ RT 10 Magnetic Stirring Hotplate
10-position digital magnetic hotplate stirrer for synchronous heating and stirring. IKA™ Stainless Steel Digital Magnetic Stirrer provides noiseless and consistent stirring on all positions.
Brand: IKA™ 0003691100
2739.48 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Speed remains constant, even when load changes.
- Homogeneous temperature distribution across the heating plate
- Adjustable reverse operation
- Eco-mode for a low self-warming of the surface
- Wear free magnetic coils
- Error code display
- Easy operation with touch keypad
- Suitable for continuous operation
- Speed range: 0 to 1000 rpm
Specifications
0 to 1000 rpm | |
100/115/230 V | |
190 mm | |
50/60 Hz | |
10 RPM Steps | |
RT 10 Magnetic Stirrer |
10 | |
410 w | |
230/115/100V, 50/60Hz, 410w |
|
EU | |
5°C to 40°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction