Learn More
Corning™ RPMI 1640, 1X
Click to view available options
Quantity:
6 x 500 mL
Product Type:
RPMI 1640, No L-glutamine, No Phenol Red
pH:
7 to 7.4
Unit Size:
Pack of 6
Description
Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol. Cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.
- Can be used for suspension cultures or monolayer human leukemic cells
- This formulation is supplemented with glutaGRO™ in place of L-glutamine for improved stability
Specifications
Specifications
| Concentration | 1 X |
| Product Type | RPMI 1640, No L-glutamine, No Phenol Red |
| Without Additives | No Glutamine, No Phenol Red |
| For Use With (Application) | To assess growth promotion and cytotoxicity characteristics |
| Quantity | 6 x 500 mL |
| Endotoxin Level | <0.25EU/mL |
| Formulation | L-Cystine/L-Methionine, Sodium Bicarbonate |
| pH | 7 to 7.4 |
| Content And Storage | 2° to 8°C |