Learn More
Fisherbrand™ Cylindrical Stir Bars with Removable Pivot Ring
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
50.8 mm
Unit Size:
Each
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | FDA, ISO 9001:2008 |
| Color | White |
| Diameter (English) | 0.31in. |
| Includes | Removable pivot ring |
| Length (English) | 2in. |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 50.8 mm |
| Material | Alnico Magnet |
| Shape | Cylinder |