Learn More
Korff™ Round Aluminum Foil Discs
€ 182.00 - € 300.00
Specifications
Material | Aluminum |
---|---|
For Use With (Application) | Used for wide variety of laboratory tasks |
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10313841
|
Korff™
60210 |
100 mm |
€ 182.00
Pack of 1000 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
10272041
|
Korff™
60214 |
120 mm |
€ 213.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
12672603
|
Korff™
60216 |
130 mm |
€ 200.00
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10343451
|
Korff™
60216 |
130 mm | N/A | ||||||
10728521
|
Korff™
60220 |
150 mm |
€ 300.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
Specifications
Aluminum | |
Used for wide variety of laboratory tasks |