Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Brand:  SVL™ RF13
112.10 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 10203180
  • € 118.00 / Pack of 5
Estimated Shipment: 13-08-2025
to see stock.
Explore more special offers
Description
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200°C
  • System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
  • Joint can be retained together with spring loaded metal clip
  • Component parts RM, RF, RO and RC should be ordered separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Female joint
Glass, PTFE
Spherical joint
13
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SVL™ Rotulex™ Spherical Joints > Bore size: 5mm; Joint size: 13