Learn More
Eppendorf™ Rotor Lids
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
A-2-MTP
Centrifuge 5427R, FA-45-24-11-Kit
Centrifuge 5427R, FA-45-30-11
F-34-6-38
F-45-17-17-Cyro
F-45-30-11
F-45-48-11
F-45-64-5-PCR
FA-45-16-17
FA-45-24-11
FA-45-24-11-HS
FA-45-24-11-Kit
FA-45-30-11
FA-45-48-11
S-24-11-AT
Material:
Aluminum
Aluminum, Coated
Polypropylene
Description:
Aerosol-tight
Aerosol-tight, QuickLock
Coated
Non-aerosol-tight
Wind shield upper shell
Unit Size:
Each
Description
- Replacement rotor lids
- Aerosol-tight and aluminum lids available
- Available in a variety of sizes
Specifications
Specifications
| Type | Rotor Lid |
| Material | Aluminum |
| For Use With (Equipment) | F-45-64-5-PCR |
| Description | Non-aerosol-tight |