Learn More
Benchmark Scientific™ RotoBot™ Rotating Mixers
RotoBot™ Rotating Mixers feature advanced programming options, digital speed control and more than 10 operating modes. A wide variety of rotating, mixing and vibrating actions can be combined to provide the precise agitation profiles. Supplied with holders for all common tubes from 1.5 to 50 mL.
Brand:  Benchmark Scientific™ R4040-E
 View more versions of this product
Product Code. 17934432
  • € 728.00 / Each
Explore more special offers
Specifications
360°
3 A
1.5 to 50 mL Tubes
29 x 13.7 x 29.9 cm
Up to 90% Non-condensing
100/240 V
Tubes from 1.5 mL to 50 mL
EU
Digital
6 x 1.5 mL Holders (2), 1 x 15 mL Holder (2), 1 x 50 mL Holder (2)